missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Water Hardness Test Kits
Specifications
Specifications
| Analytes | Phthalein complexone |
| For Use With (Application) | For measurement of Water hardness, total hardness |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Range | 0.7 to 30.1 °d, 5 to 215 mg/L Ca |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | Water Hardness, Total |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?