missing translation for 'onlineSavingsMsg'
Learn More

Water, DNase, RNase Free, MP Biomedicals™

Product Code. 11475792 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475792 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475792 Supplier MP Biomedicals Supplier No. 04821739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for use with RNase-Erase™ and DNA-Erase™ decontaminants

TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Molecular Formula H2O
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02
Quantity 500 mL
Chemical Name or Material Water, Sterile, Nuclease Free
Grade A sterile solution, completely free of detectable DNase (exo and endo) and RNase; DEPC treated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.