missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water in DMSO, 90% by volume, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
200 L
Unit Size:
200 L
Specifications
Specifications
| Color | Colorless |
| CAS | 67-68-5, 7732-18-5 |
| Quantity | 200 L |
| Molecular Formula | H2O |
| Packaging | Plastic Drum |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| SMILES | O |
| IUPAC Name | water |
| Molecular Weight (g/mol) | 18.02 |
| Concentration or Composition (by Analyte or Components) | 90% water, 10% DMSO (by volume) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction