missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Water, deionised and demineralised

Product Code. 15387526
Change view
Click to view available options
Volume (Metric):
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
15387526 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15387526 Supplier Hach Lange™ Supplier No. 27256
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Demineralised Type 2 Water.

Demineralised Type 2 WaterSpecific Conductance: ≤1 μS/cm,Silica (SiO2): ≤30 μg/L,Chloride (Cl-): ≤20 μg/L,Hardness: Negative,Consumption of KMnO4: Negative

Specifications:

  • Platform: Solution
  • Quantity: 4 L
  • Shelf Life: 60 months from production date
TRUSTED_SUSTAINABILITY

Specifications

pH Range 5.5 to 7
Volume (Metric) 4 L
Sterility Non-sterile
Type Water Testing Kit Supplies and Reagents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.