Learn More
Fistreem International™ AquaRec UV Water Recirculation System
Click to view available options
Electrical Requirements:
220/240V 50/60Hz
Unit Size:
Each
Description
- Particularly useful for providing a purified water supply in locations where on-line water supply and drains are not available.
- The self contained system is housed in an attractive cabinet with acrylic viewing panel.
- Feed water is purified via a particulate removal cartridge and ion exchange (deionizing) resin for low ionic content water.
- p\Pure water is finally passed through a powerful ultraviolet lamp system to reduce the possibility of bacterial contamination in the chamber.
- Used water from the chamber is collected into an internal 20L reservoir in the AquaRec and continuously recirculated through the cartridges and UV to ensure a constant supply of purified water to the chamber.
- Purification cartridges are fully disposable and can be quickly exchanged.
- Two purity windows in the deionizer cartridge indicate resin exhaustion by a distinctive resin color change.
- The purity windows show water quality at 1 and 10 microSiemen cm conductivity.
- The Particulate Filter removes particles to 50 micron.
Specifications
Specifications
| Width (English) | 18.1102 in. |
| Height (Metric) | 740mm |
| Electrical Requirements | 220/240V 50/60Hz |