missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™ Plus, for HPLC, Honeywell Riedel-de Haën™

Product Code. 15654670
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
Pack of 4
This item is not returnable. View return policy

Product Code. 15654670

Brand: Honeywell-Chromasolv 348774X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Formula Weight 18.02
Grade HPLC
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 4 x 2.5 L
Chemical Name or Material Water, CHROMASOLV™ Plus, for HPLC
Product Identifier
  • Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.