missing translation for 'onlineSavingsMsg'
Learn More

Water (Biotech Grade), Fisher BioReagents

Product Code. p-23703
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly CUBE
Unit Size:
20 L
4 L
This item is not returnable. View return policy

Product Code. 10091543

Brand: Fisher BioReagents BP24854

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Biotech-grade water can be used for several biological test procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Merck Index 15, 10237
Formula Weight 18.02
Grade Biotech
Packaging Poly CUBE
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Density 1.000g/cm³
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.