missing translation for 'onlineSavingsMsg'
Learn More

Water 50%, Acetonitrile 50%, LabReady™ Solvent Blend, for LC-MS and HPLC, Honeywell Burdick & Jackson

Product Code. 17854112 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Unit Size:
16 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
17854112 4 x 4 L Glass Bottle 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17854112 Supplier Honeywell-Burdick and Jackson Supplier No. LC4354
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7732-18-5, 75-05-8, 7732-18-5
CAS Min % 50.0000%, 43.9000%
CAS Max % 56.1000%, 50.0000%
Density 0.7857 g/cm3
Quantity 4 x 4 L
Flash Point 3°C
Packaging Glass Bottle
Molecular Formula C2H6O
UN Number UN1648
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 82°C
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 962
ChEBI CHEBI:15377
Grade HPLC
Chemical Name or Material Water 50%, Acetonitrile 50%, LabReady™ Solvent Blend, for LC-MS and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.