missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% TFA, LC/MS grade, Thermo Scientific
GREENER_CHOICE

Product Code. 30280963 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Unit Size:
2.5 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280963 2.5 L 2.5 L
30280961 500 mL 500mL
30280945 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280963 Supplier Thermo Scientific Supplier No. TL01192500
 Restricted Item
Estimated Shipment: 26-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Quantity 2.5 L
Form Liquid
Reagent Type Solvent Mixture
For Use With Liquid Chromatography, Mass Spectrometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.