missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% formic acid, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™

Product Code. 15634340
Change view
Click to view available options
Quantity:
1 L
4 x 4 L
Unit Size:
1 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634340 4 x 4 L Pack of 4
15614340 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15634340 Supplier Honeywell Riedel-de Haën Supplier No. 346734X4L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

MDL Number MFCD00011332

Specifications

Quantity 4 x 4 L
UN Number NONH for all modes of transport
Formula Weight 18.02g/mol
Chemical Name or Material Water with 0.1% formic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.