missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WASP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537303
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 53 kDa. Purity is >95% by SDS-PAGE.
The Wiskott-Aldrich syndrome family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. Wiskott-Aldrich syndrome is a rare, inherited, X-linked, recessive disease characterized by immune dysregulation and microthrombocytopenia, and is caused by mutations in the WAS gene. The WAS gene product is a cytoplasmic protein, expressed exclusively in hematopoietic cells, which show signalling and cytoskeletal abnormalities in WAS patients. A transcript variant arising as a result of alternative promoter usage, and containing a different 5' UTR sequence, has been described, however, its full-length nature is not known.
Specifications
| WASP | |
| Polyclonal | |
| Unconjugated | |
| Was | |
| 2900021I12Rik; 3110031I02Rik; eczema-thrombocytopenia; IMD2; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; SCNX; THC; THC1; thrombocytopenia 1 (X-linked); TRS4; U42471; Was; WASL; Wasp; WASPA; WASPB; Wiskott-Aldrich syndrome; Wiskott-Aldrich syndrome homolog; Wiskott-Aldrich syndrome homolog (human); Wiskott-Aldrich syndrome like; wiskott-Aldrich syndrome protein; Wiskott-Aldrich syndrome protein homolog; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7454 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42768 | |
| Was | |
| Recombinant full length human WASP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction