missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Washington Residual Solvent Mixture 765 10000 μg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer
Click to view available options
Quantity:
4.5 mL
Unit Size:
4.5mL
Specifications
Specifications
| CAS | 106-97-8, 74-98-6 |
| Quantity | 4.5 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | N,N-Dimethylacetamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction