missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Washing Buffer II for GeneJET Genomic DNA Purification Kits (concentrated), Thermo Scientific™
Shop All Thermo Scientific Products
Click to view available options
Quantity:
40 mL
missing translation for 'unitSize'
40mL
Beschreibung
Thermo Scientific Washing Buffer II (concentrated) is a component of the GeneJET Genomic DNA Purification Kit (K0721/K0722), GeneJET Whole Blood Genomic DNA Purification Mini Kit (K0781, K0782), and the GeneJET Plant Genomic DNA Purification Mini Kit (K0791/K0792) and may be purchased separately. Provided in 40 mL aliquots.
Spezifikation
Spezifikation
| Chemical Name or Material | Wash buffer |
| Description | Washing Buffer II For GeneJET Genomic DNA Purification Kit, 40mL, Concentrated, Liquid, Wash Buffer, Used For DNA Extraction |
| For Use With (Application) | DNA and RNA purification and analysis, DNA extraction, genomic DNA purification, gDNA from animal tissues and cells, gDNA from bacteria, gDNA from plant and fungi, gDNA from buccal cells, gDNA from blood (medium-scale, 100μL-1mL) |
| Physical Form | Liquid |
| Quantity | 40 mL |
| Formulation | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur