missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Wash Buffer RA3 Concentrate
Description
- For use with NucleoSpin RNA kits, including: RNA, RNA Plant, RNA / Protein and RNA Clean-up kits
- Wash buffers are used to wash the silica membrane filter after binding, prior to eluting the RNA
- Supplemental buffers can be purchased separately
Specifications
Specifications
| Chemical Name or Material | RA3 Concentrate |
| Color | Colorless |
| Concentration | 4X |
| For Use With (Application) | Purifying total RNA |
| For Use With (Equipment) | NucleoSpin™ RNA kits |
| pH | 7 to 8 |
| Physical Form | Liquid |
| Quantity | 25 mL |
| Odor | Odorless |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?