missing translation for 'onlineSavingsMsg'
Learn More

Wash Buffer WB1 (concentrated), Thermo Scientific™

Product Code. 15544424 Shop All Thermo Scientific Products
Click to view available options
Quantity:
40 mL
Unit Size:
40mL
This item is not returnable. View return policy

Product Code. 15544424

Brand: Thermo Scientific R1711

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wash Buffer WB1 (concentrated)

Thermo Scientific GeneJET Wash Buffer WB1 (concentrated) is a component of several GeneJET nucleic acid purification kits and may be purchased separately. Provided in 40 mL aliquots. Please consult kit protocols to confirm catalog numbers for re-ordering the appropriate kit-specific components.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Wash buffer
Color Colorless
Density 0.86g/cm3
Description Wash Buffer WB1, 40mL, Concentrated, Colorless, Solution, Wash Buffer, 0.86g/cm3 Density, Used For DNA Extraction
For Use With (Application) DNA and RNA purification and analysis, DNA extraction, genomic DNA purification, gDNA from blood (medium-scale, 100μL-1mL)
Physical Form Solution
Quantity 40 mL
Formulation Solution
Odor Odorless

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.