missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer NT3 Concentrate
Description
NucleoTraP™ and NucleoSpin™ PCR clean-up kits allow DNA fragments to be purified from enzymatic reactions, such as PCR, as well as from agarose gels. Compatible kits include:
- NucleoSpin™ Gel and PCR Clean-up (standard, Midi, Maxi)
- NucleoSpin™ 8/96 PCR Clean-up (full and core kit)
- NucleoTraP™ CR and NucleoTraP™
Specifications
Specifications
| Chemical Name or Material | Wash buffer |
| Color | Colorless |
| Concentration | 5X (50 mL concentrate yields 125 mL working solution) |
| For Use With (Application) | Purification (clean up) of DNA |
| For Use With (Equipment) | NucleoTraP™ and NucleoSpin™ PCR clean-up kits |
| pH | 7.0 to 8.0 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Odor | Odorless |
| Packaging | Concentrated |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?