Learn More
Macherey-Nagel™ Wash Buffer N4-EF
Product Code. 30694925
Change view
Click to view available options
Quantity:
3 L
10 L
Unit Size:
10 L
3 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30694925 10 L 10 L
30694926 3 L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30694925 Supplier Macherey-Nagel™ Supplier No. 740795.10
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Wash Buffer N4‐EF to be used with NucleoBond PC EF kits
  • For NucleoBond PC EF kits
Specifications
For Use With (Application) Endotoxin-free Plasmid DNA Isolation
Quantity 10 L
Solution Type Buffer