missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Wash Buffer AW

Product Code. 12706563
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12706563

Brand: Macherey-Nagel™ 740916.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for bacterial strains with high endogenous nuclease activity or for improvement of sequencing results. Macherey-Nagel™ NucleoSpin™ Wash Buffer AW is used with NucleoSpin™ Plasmid DNA purification kits for rapid small-scale preparation of high-copy plasmids from E. coli.

The NucleoSpin™ Plasmid kit is designed for purifying up to 20μg of high copy-number plasmid DNA from 1mL to 5mL E. coli overnight cultures preferably grown in LB medium. The NucleoSpin™ Plasmid binding strips contain a unique silica membrane that binds DNA in the presence of chaotropic salt.

After a modified alkaline lysis procedure, bacterial lysates are loaded onto the NucleoSpin™ Plasmid filter strips arranged on the NucleoVac 96 vacuum manifold. Filtrates are collected directly in the NucleoSpin™ Plasmid binding strips. After binding and washing, highly pure plasmid DNA is eluted in low-salt buffer (5mM Tris/HCl, pH 8.5). Alternatively, water (pH 8.5) can be used.

Kit components: NucleoSpin™ Plasmid filter strips, NucleoSpin™ Plasmid binding strips, all necessary buffers, RNase A, MN wash plate, MN tube strips, cap strips.

  • Silica-membrane technology
  • Yield: up to 20μL sequencing grade plasmid DNA
  • Elution volume: 75μL to 150μL
  • Culture volume: 1mL to 5mL
  • Binding capacity: 25μg
  • Maximum plasmid size <15kb
  • No use of organic solvents
  • NucleoSpin 8 filter strips for convenient filtration of bacterial lysates included

For further information on this product contact your local Customer Services.

Specifications

Chemical Name or Material Wash AW Buffer
Color Colorless
For Use With (Application) For Plasmid DNA Purification
For Use With (Equipment) NucleoSpin™ Plasmid DNA Purification Kits
pH 7 to 8
Physical Form Solution
Quantity 1000 mL
Formulation Guanidinium hydrochloride: 36 to <50%; 2-Propanol: 20 to <35%
Odor Alcohol
compliance-icons compliance-icons
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.