missing translation for 'onlineSavingsMsg'
Learn More

WASF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£143.00 - £251.00

Specifications

Antigen WASF2
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18651607
View Documents
Novus Biologicals
NBP3-12675-100ul
100 μg
£251.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18674557
View Documents
Novus Biologicals
NBP3-12675-50ul
50 μg
£143.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WASF2 Polyclonal antibody specifically detects WASF2 in Human samples. It is validated for Western Blot
Specifications

Specifications

WASF2
Western Blot
Unconjugated
Rabbit
Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction
PBS, pH7.0
10163
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
IMD2, Protein WAVE-2, SCAR2suppressor of cyclic-AMP receptor (WASP-family), Verprolin homology domain-containing protein 2, WAS protein family, member 2, WASP family Verprolin-homologous protein 2, WAVE2dJ393P12.2, wiskWASP family protein member 2
Produced in rabbits immunized with E. coli-derived Human WASF2 fragment
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.