missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASF1/WAVE1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | WASF1/WAVE1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
WASF1/WAVE1 Polyclonal antibody specifically detects WASF1/WAVE1 in Human samples. It is validated for Western BlotSpecifications
| WASF1/WAVE1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KIAA0269WAVEhomology of dictyostelium scar 1, Protein WAVE-1, SCAR1WASP family protein member 1, Verprolin homology domain-containing protein 1, WAS protein family, member 1, WAVE1FLJ31482, wisk | |
| A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human WASF1 (NP_001020107.1). VARAAPVCETVPVHPLPQGEVQGLPPPPPPPPLPPPGIRPSSPVTVTALAHPPSGLHPTPSTAPGPHVPLMPPSPPSQVIPASEPKRHPSTLPVISDARSVLLEAIRKGIQLRKVEEQREQEAKHERIENDVATILSRRIAVEYSDSEDDSEFDEVDWLE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8936 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title