missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567064
This item is not returnable.
View return policy
Description
Immunogen sequence: MELVQDLAQGF NKKYGEFFPV PESILTSMKK VKSLRDPSAK MSKSDPDKLA TVRITDSPEE IVQKFRKAVT DFTSEVTYDP A Highest antigen sequence identity to the following orthologs - mouse 90%, rat 88%.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.
Specifications
| WARS2 | |
| Polyclonal | |
| Unconjugated | |
| WARS2 | |
| (Mt)TrpRS; 5730427B17Rik; 9430020O07Rik; AI413375; mitochondrial rryptophanyl-tRNA synthetase-like protein; mitochondrial tryptophanyl-tRNA synthetase 2; trpRS; tryptophan tRNA ligase 2, mitochondrial; tryptophan--tRNA ligase, mitochondrial; tryptophanyl tRNA synthetase 2 (mitochondrial); tryptophanyl tRNA synthetase 2, mitochondrial; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, mitochondrial; tryptophanyl-tRNA synthetase, mitochondrial; tryptophan--tRNA ligase, mitochondrial; WARS2; zgc:110828 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGM6 | |
| WARS2 | |
| Recombinant Human WARS2. Recombinant protein control fragment (Product #RP-107704). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction