missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529849
This item is not returnable.
View return policy
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (87%), Xenopus laevis (88%), Rabbit (96%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. Tryptophanyl-tRNA synthetase (WARS) catalyzes the aminoacylation of tRNA(trp) with tryptophan and is induced by interferon. Tryptophanyl-tRNA synthetase belongs to the class I tRNA synthetase family. Four transcript variants encoding two different isoforms have been found for this gene.
Specifications
| WARS | |
| Polyclonal | |
| Unconjugated | |
| WARS1 | |
| GAMMA-2; hWRS; IFI 53; IFI53; IFP 53; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, cytoplasmic; Wars; WARS protein; Wrs | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22375, 7453 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol; pH 7 | |
| P23381, P32921 | |
| WARS1 | |
| Recombinant protein encompassing a sequence within the center region of human Tryptophanyl tRNA synthetase. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction