missing translation for 'onlineSavingsMsg'
Learn More

Warfarin EP Impurity C, Mikromol™

Product Code. 30459718
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30459718

Brand: Mikromol MM0732.0300254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Warfarin EP Impurity C, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzalacetone
CAS 1896-62-4
Analyte or Component Names Benzalacetone
Shipping Condition Room Temperature
Molecular Formula C10 H10 O
Certifications/Compliance ISO 9001
Synonym (3E)-4-Phenylbut-3-en-2-one (Benzalacetone), (3E)-4-Phenylbut-3-en-2-one, Warfarin Sodium Imp. C (EP), Benzalacetone, Warfarin Sodium Impurity C, Warfarin Impurity C
InChI Formula InChI=1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+
Solution Type Neat
SMILES CC(=O)\C=C\c1ccccc1
IUPAC Name (E)-4-phenylbut-3-en-2-one
Molecular Weight (g/mol) 146.19
Quantity 4 x 25 mg
Formula Weight 146.0732 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.