missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Warfarin EP Impurity C, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Warfarin EP Impurity C
Specifications
Specifications
| Chemical Name or Material | Benzalacetone |
| CAS | 1896-62-4 |
| Analyte or Component Names | Benzalacetone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H10 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3E)-4-Phenylbut-3-en-2-one (Benzalacetone), (3E)-4-Phenylbut-3-en-2-one, Warfarin Sodium Imp. C (EP), Benzalacetone, Warfarin Sodium Impurity C, Warfarin Impurity C |
| InChI Formula | InChI=1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+ |
| SMILES | CC(=O)\C=C\c1ccccc1 |
| IUPAC Name | (E)-4-phenylbut-3-en-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction