missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WAPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530458
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Mouse (95%), Chicken (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
Specifications
| WAPL | |
| Polyclonal | |
| Unconjugated | |
| WAPL | |
| A530089A20Rik; BC037674; DIF-2; Dioxin-inducible factor 2; FOE; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); friend of EBNA2 protein; KIAA0261; RGD1308960; WAPAL; WAPL; WAPL cohesin release factor; wings apart-like homolog (Drosophila); wings apart-like protein homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.72 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z5K2 | |
| WAPL | |
| Recombinant protein encompassing a sequence within the center region of human WAPL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction