missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAPL Antibody (SR1232), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22099-50ul
This item is not returnable.
View return policy
Description
WAPL Monoclonal antibody specifically detects WAPL in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| WAPL | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| SR1232 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
| A synthesized peptide derived from human WAPL (Uniprot #: Q7Z5K2) | |
| 50 μg | |
| DNA Repair | |
| 23063 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction