missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAPL Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81779AF350
This item is not returnable.
View return policy
Description
WAPL Polyclonal antibody specifically detects WAPL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WAPL | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
| WAPL antibody was raised against an 18 amino acid peptide near the amino terminus of human WAPL. The immunogen is located within amino acids 30 - 80 of WAPL. Amino Acid Squence: LGQKRPNFKPDIQEIPKK | |
| 0.1 mL | |
| DNA Repair | |
| 23063 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction