missing translation for 'onlineSavingsMsg'
Learn More

Wang resin, 1% cross-linked, 0.8-1.1mmol/g, 200-400 mesh, Thermo Scientific™

Product Code. 11373926
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373926 25 g 25g
11363926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373926 Supplier Thermo Scientific Alfa Aesar Supplier No. L17028.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201058-08-4
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00131778
InChI Key NERFNHBZJXXFGY-UHFFFAOYSA-N
Synonym 4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
PubChem CID 2734007
SMILES CC1=CC=C(COC2=CC=C(CO)C=C2)C=C1

Specifications

Mesh Size 200-400 Mesh
Quantity 25 g
IUPAC Name {4-[(4-methylphenyl)methoxy]phenyl}methanol
Chemical Name or Material Wang resin, 1% cross-linked
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.