missing translation for 'onlineSavingsMsg'
Learn More

Walrycin B, MedChemExpress

Product Code. 16453507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453507 50 mg 50mg
16419967 5 mg 5mg
16407488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16453507 Supplier Medchem Express Supplier No. HY18219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Walrycin B is a novel antibacterial compound specifically targeting the essential WalR response regulator.

Walrycin B is a novel antibacterial compound specifically targeting the essential WalR response regulator.

Specifications

Chemical Name or Material Walrycin B
Physical Form Solid
CAS 878419-78-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H10F3N5O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 11.5 mg/mL (34.10 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC(C2=CC=C(C(F)(F)F)C=C2)=NN(C)C1=N3)N(C)C3=O
Molecular Weight (g/mol) 337.26
Formula Weight 337.26
Percent Purity 97.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.