missing translation for 'onlineSavingsMsg'
Learn More

BICASA Wall Bench MECC Series with Sink

Product Code. 30057591 Shop All BICASA Products
Change view
Click to view available options
Dimensions (L x W x H):
600 x 900 x 930 mm
1200 x 900 x 930 mm
600 x 900 x 2100 mm
1200 x 900 x 2100 mm
Color:
Anthracite
Blue
Red
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Color unitSize
30057591 600 x 900 x 930 mm Anthracite Each
30057556 600 x 900 x 930 mm Blue Each
30057608 600 x 900 x 930 mm Red Each
30057539 600 x 900 x 2100 mm Anthracite Each
30057590 600 x 900 x 2100 mm Blue Each
30057548 600 x 900 x 2100 mm Red Each
30057573 1200 x 900 x 930 mm Anthracite Each
30057550 1200 x 900 x 930 mm Blue Each
30057574 1200 x 900 x 930 mm Red Each
30057559 1200 x 900 x 2100 mm Anthracite Each
30057544 1200 x 900 x 2100 mm Blue Each
30057586 1200 x 900 x 2100 mm Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 30057591 Supplier BICASA Supplier No. MS.0600.900.930.PPA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wall Bench MECC Series with Sink, complete with lateral pedestals, MECC service spine, C-Frame side support, cross-bar, PP sink countertop with 1 basin, water mixer, eyewash and pegboard included.

  • Laboratory benches MECC Series
  • PEDESTALS: Aluminum drawn profile (150 x 25 mm and 150 x 50 mm) with vertical slots
  • SERVICE SPINE: Modular steel frame assembled to the pedestals at different heights with utility panels horizontal rows
  • Trunking structure with passageway for piping and wirings
  • UTILITY STRIP: predrilled metal band at countertop height
  • UTILITY PANELS: Moulded Polypropylene PP resin cover (V0 flame retarded - IEC 60695-11-10 / -20)
  • C-FRAME WORKSTATIONS: certified according to EN 13150:2001 and BRG 234/09.06
  • C-shaped steel tubular frame levelling supports - Horizontal cross-bars - Lateral metal sheeting
  • COUNTERTOPS: Independent units - installed and fixed above the C-FRAME
  • COLORS: METAL: Light Grey matt bonderized Polyurethane powder coating
  • UTILITY MECC PANELS: Silver Grey (frontal plate) - ABS Electric panel rear box: Black
  • FIRE-RETARDANT PARTICLEBOARD 19 mm panels: Light Grey (2 decorative)
  • TAPS: On turret - Epoxy powder coating - Light Grey color
  • Locking on utility strip or on countertop with threaded inlet feed sleeve and lock nut
  • Fluid identification (handle according to EN13792:2000)
  • REAGENT HOLDER SHELF: coated metal - cantilever
  • PEGBOARD: Plastic (acrylic stand) panel

Specifications

Product Type Wall Bench
Color Anthracite
Dimensions (L x W x H) 600 x 900 x 930 mm
Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.