missing translation for 'onlineSavingsMsg'
Learn More

W-54011, MedChemExpress

Product Code. 16447248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447248 10 mg 10mg
16441659 1 mg 1mg
16419777 5 mg 5mg
16413267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447248 Supplier Medchem Express Supplier No. HY16992A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress W-54011 is a potent and orally active non-peptide C5a receptor antagonist. W-54011 inhibits the binding of 125I-labeled C5a to human neutrophils with a Ki value of 2.2 nM. W-54011 also inhibits C5a-induced intracellular Ca2+ mobilization, chemotaxis, and generation of ROS in human neutrophils with IC50s of 3.1 nM, 2.7 nM, and 1.6 nM, respectively.

W-54011 is a potent and orally active non-peptide C5a receptor antagonist. W-54011 inhibits the binding of 125I-labeled C5a to human neutrophils with a Ki value of 2.2 nM. W-54011 also inhibits C5a-induced intracellular Ca2+ mobilization, chemotaxis, and generation of ROS in human neutrophils with IC50s of 3.1 nM, 2.7 nM, and 1.6 nM, respectively.

Specifications

Chemical Name or Material W-54011
CAS 405098-33-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H37ClN2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 28 mg/mL (56.79 mM)
SMILES O=C(C1CCCC2=C1C=C(OC)C=C2)N(CC3=CC=C(N(C)C)C=C3)C4=CC=C(C(C)C)C=C4.[H]Cl
Molecular Weight (g/mol) 493.08
Formula Weight 493.08
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.