missing translation for 'onlineSavingsMsg'
Learn More

W-5 hydrochloride, Tocris Bioscience™

Product Code. 18055232
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18055232 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055232 Supplier Tocris Bioscience Supplier No. 0368/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calmodulin antagonist

Calmodulin antagonist (inhibits Ca2+-calmodulin dependent PDE with an IC50 of 240 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 61714-25-8
Molecular Formula C16H23ClN2O2S
MDL Number MFCD00058043
Quantity 10 mg
Synonym w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde
InChI Key HOCSVIGHWPLMFC-UHFFFAOYSA-N
SMILES [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1
IUPAC Name hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride
Molecular Weight (g/mol) 342.88
PubChem CID 173829
Formula Weight 342.88
Percent Purity >98%
Chemical Name or Material W-5 hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.