missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VZ 185

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Potent and selective VHL-based dual Degrader (PROTAC™) of BRD7/9 (DC50 values are 4 and 34 nM for BRD7 and BRD9, respectively). Exhibits selectivity for BRD7/9 degradation over other bromodomain-containing proteins and other BAF/PBAF subunits. Displays cytotoxicity towards EOL-1 and A-204 cancer cell lines.
Negative control cis VZ 185 (Cat. No. 6939) also available.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-N-(2-((5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyl)oxy)-4-(4-methylthiazol-5-yl)benzyl)-1-((S)-2-(1-fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide |
| CAS | 2306193-61-1 |
| Quantity | 5 mg |
| Molecular Formula | C53H67FN8O8S |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction