missing translation for 'onlineSavingsMsg'
Learn More

VX-984, MedChemExpress

Product Code. 16450438
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450438

Brand: Medchem Express HY19939S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.

VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.

Specifications

Chemical Name or Material VX-984
CAS 1476074-39-1
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C23H21D2N7O
Color White
Synonym M9831
Solubility Information DMSO : 10 mg/mL (24.07 mM; Need ultrasonic)
SMILES O=C(C1=CC=NC2=C([C@H](C)CNC3=CC(C4=C([2H])N=C(C)N=C4[2H])=NC=N3)C=CC=C12)NC
Industry Type Cancer-Kinase/protease
Molecular Weight (g/mol) 415.49
Quantity 5 mg
Formula Weight 415.49
Percent Purity 98.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.