missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VWCE Polyclonal Antibody
GREENER_CHOICE

Product Code. 15758381 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15758381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15758381 Supplier Invitrogen™ Supplier No. PA560551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GGDECTTCVC QNGEVECSFM PCPELACPRE EWRLGPGQCC FTCQEPTPST GCSLDDNGVE FPIGQIWSPG DPCELCICQA DGSVSCKRTD CVDSCP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VWCE
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VWCE
Gene Accession No. Q96DN2
Gene Alias 1300015B04Rik; HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; RGD1309910; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; Vwce
Gene Symbols VWCE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VWCE. Recombinant protein control fragment (Product #RP-97456).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.