missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWA8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558648
This item is not returnable.
View return policy
Description
Immunogen sequence: EHAIKEIVKE EADEYFVIVL SDANLSRYGI HPAKFAQILT RDPQVNAFAI FIGSLGDQAT RLQRTLPAGR SFVAMDTKDI PQILQQIFTS TML Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 25%.
Specifications
| VWA8 | |
| Polyclonal | |
| Unconjugated | |
| VWA8 | |
| KIAA0564; P7BP2; PEX7-binding protein 2; von Willebrand factor A domain containing 8; von Willebrand factor A domain-containing protein 8; VWA8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23078 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A3KMH1 | |
| VWA8 | |
| Recombinant protein corresponding to Human VWA8. Recombinant protein control fragment (Product #RP-97927). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction