Learn More
VWA5B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566723
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: CNIISKYTAFV PVDVSKSRYL PTVVEYPNSG AALRMLGSRA LAQQWRGTSS GFGRPQTMLG EDSAPGN Highest antigen sequence identity to the following orthologs - mouse 62%, rat 57%.
VWA5B1 is a protein coding gene.Specifications
VWA5B1 | |
Polyclonal | |
Unconjugated | |
VWA5B1 | |
von Willebrand factor A domain containing 5B1; von Willebrand factor A domain-containing protein 5B1; VWA5B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127731 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TIE3 | |
VWA5B1 | |
Recombinant Human VWA5B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only