missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWA5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530082
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in tumorigenesis as a tumor suppressor. Altered expression of this protein and disruption of the molecular pathway it is involved in, may contribute directly to or modify tumorigenesis.
Specifications
| VWA5A | |
| Polyclonal | |
| Unconjugated | |
| Vwa5a | |
| 5830475I06Rik; AW552491; BCSC1; BCSC-1; breast cancer suppressor candidate 1; E130119J22; Loh11cr2a; loss of heterozygosity 11 chromosomal region 2 gene A protein; Loss of heterozygosity 11 chromosomal region 2 gene A protein homolog; loss of heterozygosity, 11, chromosomal region 2, gene A; loss of heterozygosity, 11, chromosomal region 2, gene A homolog; Masa1; masa-1; Mast cell surface antigen 1; mast cell surface antigen-1; ortholog of mouse AW551984; von Willebrand factor A domain containing 5A; von Willebrand factor A domain-containing protein 5A; VWA5A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O00534 | |
| Vwa5a | |
| Recombinant protein encompassing a sequence within the center region of human VWA5A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction