Learn More
VWA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14322-1-AP-150UL
 View more versions of this product
Product Code. 16842904
  • £349.00 / 150µL
Description
VWA1 (also known as WARP) is a member of the von Willebrand factor A-domain (VA-domain) superfamily of extracellular matrix proteins which may play a role in cartilage structure and function. VWA1 is a multimeric component of the chondrocyte pericellular matrix in articular cartilage and intervertebral disc, where it interacts with the basement membrane heparan sulfate proteoglycan perlecan. VWA1 may have a role in maintaining the blood-brain barrier.
Specifications
Specifications
VWA1
Polyclonal
Unconjugated
VWA1
VWA 1, VWA1, WARP
Rabbit
Antigen Affinity Chromatography
RUO
246228, 298683, 64856
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q642A6, Q6PCB0, Q8R2Z5
VWA1
VWA1 Fusion Protein Ag5565
150 μL
Primary
Human, Mouse, Rat