missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWA1 Monoclonal Antibody (559915)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561636
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
VWA1 (also known as WARP) is a member of the von Willebrand factor A-domain (VA-domain) superfamily of extracellular matrix proteins which may play a role in cartilage structure and function. VWA1 is a multimeric component of the chondrocyte pericellular matrix in articular cartilage and intervertebral disc, where it interacts with the basement membrane heparan sulfate proteoglycan perlecan. VWA1 may have a role in maintaining the blood-brain barrier.
Specifications
| VWA1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6PCB0 | |
| Vwa1 | |
| Mouse myeloma cell line NS0-derived recombinant human WARP, Arg19-Pro445. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 559915 | |
| Unconjugated | |
| Vwa1 | |
| 4932416A11Rik; RGD1311476; von Willebrand factor A domain containing 1; von Willebrand factor A domain-containing protein 1; von Willebrand factor A domain-related protein; Vwa1; Warp | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 64856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction