missing translation for 'onlineSavingsMsg'
Learn More

VU6005649, MedChemExpress

Product Code. 16465126
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465126

Brand: Medchem Express HY107982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU6005649 is a CNS penetrant mGlu7/8 receptor agonist with EC50s of 0.65 μM and 2.6 μM for mGlu7 receptor and mGlu8 receptor, respectively.

VU6005649 is a CNS penetrant mGlu7/8 receptor agonist with EC50s of 0.65 μM and 2.6 μM for mGlu7 receptor and mGlu8 receptor, respectively.

Specifications

Chemical Name or Material VU6005649
CAS 2137047-43-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12F5N3O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (139.95 mM; Need ultrasonic)
SMILES CC1=NN2C(N=C(C)C=C2C(F)(F)F)=C1C3=CC=C(OC)C(F)=C3F
Molecular Weight (g/mol) 357.28
Formula Weight 357.28
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.