missing translation for 'onlineSavingsMsg'
Learn More

VU0463271, MedChemExpress

Product Code. 16427077
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16427077

Brand: Medchem Express HY110110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU0463271 is a selective KCC2 antagonist, with an IC50 of 61 nM.

VU0463271 is a selective KCC2 antagonist, with an IC50 of 61 nM.

Specifications

Chemical Name or Material VU0463271
Physical Form Powder
CAS 1391737-01-1
Color Pink
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H18N4OS2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 25 mg/mL (65.36 mM; Need ultrasonic)
SMILES O=C(N(C1CC1)C2=NC(C)=CS2)CSC3=NN=C(C4=CC=CC=C4)C=C3
Molecular Weight (g/mol) 382.5
Formula Weight 382.5
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.