missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Vu-Union Vespel/Graphite Ferrules

Use with Vacuum Vu-Union connectors.

Brand:  Restek™ 20423

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13071770

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Material: 60% Vespel/40% graphite
  • Maximum Operating Temperature: 400°C
Specifications

Specifications

Vacuum Vu-Union Connector, Fits Column <0.22 I.D., <0.4 O.D.
0.011 in.
752°F
40% Graphite, 60% Vespel
0.03 cm
0.15 cm
400°C
0.0625 in.
Ferrule
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers