missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0467485

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VU 0467485 is a potent and selective positive allosteric modulator of M4 receptor; potentiates activity of ACh at M4 (EC50 values are 26.6 and 78.8 nM at rat and human M4 receptors, respectively). Exhibits selectivity for M4 over human and rat M1-3, 5, and a panel of 200 other pharmacological targets. Displays efficacy in a rat model of antipsychotic activity. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 5-Amino-N-[(3-fluoro-4-methoxyphenyl)methyl]-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide |
| CAS | 1451994-10-7 |
| Quantity | 10 mg |
| Molecular Formula | C17H17FN4O2S |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction