missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0467485
GREENER_CHOICE

Product Code. 18798616
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18798616

Brand: Tocris Bioscience™ 6932/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective positive allosteric modulator of M4 receptors; orally bioavailable and brain penetrant

VU 0467485 is a potent and selective positive allosteric modulator of M4 receptor; potentiates activity of ACh at M4 (EC50 values are 26.6 and 78.8 nM at rat and human M4 receptors, respectively). Exhibits selectivity for M4 over human and rat M1-3, 5, and a panel of 200 other pharmacological targets. Displays efficacy in a rat model of antipsychotic activity. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-N-[(3-fluoro-4-methoxyphenyl)methyl]-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide
CAS 1451994-10-7
Quantity 10 mg
Molecular Formula C17H17FN4O2S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.