missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0463271
GREENER_CHOICE

Product Code. 18774049
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18774049

Brand: Tocris Bioscience™ 4719/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective KCC2 inhibitor

VU 0463271 is a potent and selective inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 61 nM); displays >100-fold selectivity versus the Na-K-2Cl cotransporter 1 (NKCC1) and no activity against a panel of 68 GPCRs, ion channels and transporters.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide
CAS 1391737-01-1
Quantity 10 mg
Target KCC2 Inhibitors
Molecular Formula C19H18N4OS2
Formula Weight 382.5
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.