missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0360172 hydrochloride
GREENER_CHOICE

Product Code. 30001483
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001483

Brand: Tocris Bioscience™ 4323/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of mGlu5 receptors

Positive allosteric modulator of mGlu5 receptors (EC50 = 16 nM; Ki = 195 nM). Selective for mGlu5; displays no significant activity at mGlu1, mGlu2 or mGlu4 receptors. Exhibits antipsychotic-like activity in a rodent model. Also reduces spontaneous spike and wave discharges without affecting motor behaviour in a rat model of absence epilepsy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309976-62-2
Molecular Formula C18H16ClFN2O

Specifications

Quantity 10 mg
Formula Weight 330.78
Purity >98%
Molecular Weight (g/mol) 330.787
Chemical Name or Material VU 0360172 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.