missing translation for 'onlineSavingsMsg'
Learn More

VU 0357121, MedChemExpress

Product Code. p-200097520
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16416488

Brand: Medchem Express HY15393

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. VU 0357121 is inactive or very weakly antagonizing at other mGlu receptor subtypes.

VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. VU 0357121 is inactive or very weakly antagonizing at other mGlu receptor subtypes.

Specifications

Chemical Name or Material VU 0357121
CAS 433967-28-3
Color White
Purity Grade Notes Research
Molecular Formula C17H17F2NO2
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (163.76 mM)
SMILES O=C(NC1=CC=C(F)C=C1F)C2=CC=C(OCCCC)C=C2
Molecular Weight (g/mol) 305.32
Formula Weight 305.32
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.