missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0255035

Description
VU 0255035 is a highly selective muscarinic M1 antagonist (Ki = 14.87 nM). Targets the M1 orthosteric site with greater than 75-fold selectivity over the other receptor subtypes. Reduces pilocarpine-induced seizures in mice. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | N-[3-Oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide |
| CAS | 1135243-19-4 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C18H20N6O3S2 |
| Formula Weight | Observed MW: 432.52 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?