missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0238429
GREENER_CHOICE

Product Code. 18043932
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18043932

Brand: Tocris Bioscience™ 3634/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective positive allosteric modulator of M5 receptors

VU 0238429 is a selective positive allosteric modulator of M5 receptors (EC50 values are 1.16, >30 and >30 μM at M5, M1 and M3 receptors respectively). Displays no activity at M2 and M4 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.97
Content And Storage Store at -20°C
Quantity 10 mg
Product Type VU 0238429
Molecular Weight (g/mol) 351.28
Inhibitors VU 0238429

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.